Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032069.2 | NP_001027240.1 | 2 - 3 | 1005 | 94 | |
NM_078817.4 | NP_523541.2 | 2 - 3 | 1005 | 94 | ||
GenBank mRNA | BT010016.1 | - | 2 - 3 | 1004 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |