Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259165.1 | NP_001246094.1 | 13 - 14 | 2130 | 88 | |
NM_001273665.1 | NP_001260594.1 | 16 - 17 | 3199 | 88 | ||
NM_001273666.1 | NP_001260595.1 | 14 - 15 | 3412 | 88 | ||
NM_001273667.1 | NP_001260596.1 | 14 - 15 | 2557 | 88 | ||
NM_001273668.1 | NP_001260597.1 | 15 - 16 | 2761 | 88 | ||
NM_078880.3 | NP_523604.2 | 13 - 14 | 2381 | 88 | ||
NM_165313.4 | NP_724220.2 | 14 - 15 | 2723 | 88 |
Gene Symbol: CG46244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316402.1 | NP_001303331.1 | 14 - 15 | 3412 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |