Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp9b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078922.4 | NP_523646.1 | 3 - 4 | 1400 | 66 | |
GenBank mRNA | AY059439.1 | - | 3 - 4 | 1602 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |