Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078926.3 | NP_523650.2 | 6 - 7 | 804 | 60 | |
GenBank mRNA | BT023089.1 | - | 6 - 7 | 804 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster |