Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043066.1 | NP_001036531.1 | 7 - 8 | 1306 | 79 | |
NM_001299229.1 | NP_001286158.1 | 7 - 8 | 1318 | 79 | ||
NM_078926.3 | NP_523650.2 | 8 - 9 | 1240 | 79 | ||
GenBank mRNA | BT023089.1 | - | 8 - 9 | 1240 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Drosophila melanogaster |