Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274007.2 | NP_001260936.1 | 4 - 5 | 568 | 71 | |
NM_001274008.2 | NP_001260937.1 | 4 - 5 | 687 | 71 | ||
NM_001274009.2 | NP_001260938.1 | 4 - 5 | 590 | 71 | ||
NM_001274010.2 | NP_001260939.1 | 4 - 5 | 651 | 71 | ||
NM_001274011.2 | NP_001260940.1 | 4 - 5 | 573 | 71 | ||
NM_079003.5 | NP_523727.2 | 4 - 5 | 585 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |