Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079093.4 | NP_523817.1 | 2 - 3 | 586 | 57 | |
GenBank mRNA | BT125830.1 | - | 2 - 3 | 512 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |