Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsb-n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079138.2 | NP_523862.1 | 3 - 4 | 807 | 78 | |
GenBank mRNA | AY071593.1 | - | 3 - 4 | 804 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |