Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AlaRS-m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079208.3 | NP_523932.2 | 9 - 10 | 2727 | 90 | |
GenBank mRNA | AY128439.1 | - | 9 - 10 | 2680 | 90 | |
BT001744.1 | - | 9 - 10 | 2684 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |