Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079309.4 | NP_524033.2 | 3 - 4 | 1277 | 75 | |
GenBank mRNA | AY061440.1 | - | 3 - 4 | 1283 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |