Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp69D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079324.3 | NP_524048.2 | 1 - 2 | 307 | 73 | |
NM_176324.2 | NP_788502.1 | 1 - 2 | 307 | 73 | ||
GenBank mRNA | BT001531.1 | - | 1 - 2 | 303 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |