Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014588.2 | NP_001014588.1 | 2 - 3 | 622 | 100 | |
NM_079390.3 | NP_524114.1 | 2 - 3 | 447 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |