Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lush | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169997.1 | NP_001163468.1 | 2 - 3 | 431 | 72 | |
NM_079438.3 | NP_524162.1 | 2 - 3 | 444 | 72 | ||
GenBank mRNA | AF001621.1 | - | 2 - 3 | 444 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |