Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259374.2 | NP_001246303.1 | 2 - 3 | 615 | 75 | |
NM_001259375.2 | NP_001246304.1 | 2 - 3 | 1006 | 75 | ||
NM_137058.4 | NP_610902.1 | 2 - 3 | 1006 | 75 | ||
GenBank mRNA | AY069830.1 | - | 2 - 3 | 1326 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |