Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258473.1 | NP_001245402.1 | 4 - 5 | 2273 | 68 | |
NM_079878.5 | NP_524617.3 | 4 - 5 | 2023 | 68 | ||
GenBank mRNA | BT133159.1 | - | 4 - 5 | 1952 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |