Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lbk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137247.3 | NP_611091.2 | 4 - 5 | 2257 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |