Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nopo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274142.1 | NP_001261071.1 | 1 - 2 | 343 | 79 | |
NM_137461.4 | NP_611305.1 | 1 - 2 | 222 | 79 | ||
GenBank mRNA | AY060610.1 | - | 1 - 2 | 189 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |