Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038756.2 | NP_001033845.1 | 3 - 4 | 1067 | 72 | |
GenBank mRNA | BT021240.1 | - | 3 - 4 | 1065 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |