Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mur2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297860.1 | NP_001284789.1 | 8 - 9 | 6520 | 100 | |
NM_130571.3 | NP_569927.1 | 7 - 8 | 6159 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |