Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258572.1 | NP_001245501.1 | 6 - 7 | 1908 | 83 | |
NM_001297897.1 | NP_001284826.1 | 6 - 7 | 1582 | 83 | ||
NM_001297898.1 | NP_001284827.1 | 7 - 8 | 1490 | 83 | ||
NM_130651.3 | NP_570007.2 | 6 - 7 | 1582 | 83 | ||
GenBank mRNA | AY661807.1 | - | 5 - 6 | 1240 | 83 | |
BT022705.1 | - | 6 - 7 | 1585 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |