Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299917.1 | NP_001286846.1 | 4 - 5 | 1418 | 82 | |
NM_138090.2 | NP_611934.1 | 4 - 5 | 1257 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |