Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274331.1 | NP_001261260.1 | 10 - 11 | 3061 | 61 | |
NM_167889.2 | NP_728621.1 | 11 - 12 | 3184 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |