Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: promL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274422.1 | NP_001261351.1 | 12 - 13 | 3031 | 59 | |
NM_139513.3 | NP_647770.1 | 12 - 13 | 2955 | 59 | ||
NM_167986.1 | NP_728809.1 | 12 - 13 | 3233 | 59 | ||
NM_167987.4 | NP_728810.1 | 13 - 14 | 3114 | 59 | ||
GenBank mRNA | AF127935.2 | - | 13 - 14 | 3089 | 59 | |
AF197345.1 | - | 12 - 13 | 2911 | 59 | ||
BT031128.1 | - | 12 - 13 | 2913 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |