Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274543.1 | NP_001261472.1 | 4 - 5 | 1953 | 73 | |
NM_058020.4 | NP_477368.1 | 4 - 5 | 2035 | 73 | ||
NM_168160.2 | NP_729154.1 | 4 - 5 | 2032 | 73 | ||
NM_168161.2 | NP_729155.1 | 4 - 5 | 1994 | 73 | ||
GenBank mRNA | BT150282.1 | - | 4 - 5 | 2031 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |