Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168162.2 | NP_729159.2 | 10 - 11 | 2028 | 65 | |
GenBank mRNA | BT099579.1 | - | 10 - 11 | 2022 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |