Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272429.2 | NP_001259358.1 | 4 - 5 | 843 | 51 | |
NM_132277.3 | NP_572505.1 | 6 - 7 | 1937 | 51 | ||
GenBank mRNA | AB034690.1 | - | 6 - 7 | 1908 | 51 | |
AF181631.2 | - | 6 - 7 | 1909 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |