Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274336.1 | NP_001261265.1 | 1 - 2 | 318 | 65 | |
NM_139367.4 | NP_647624.1 | 2 - 3 | 777 | 65 | ||
GenBank mRNA | AY069290.1 | - | 1 - 2 | 318 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |