Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169916.2 | NP_001163387.2 | 11 - 12 | 5125 | 70 | |
NM_001202142.2 | NP_001189071.1 | 9 - 10 | 1708 | 70 | ||
NM_001202143.2 | NP_001189072.1 | 9 - 10 | 1708 | 70 | ||
NM_001259753.2 | NP_001246682.1 | 8 - 9 | 1387 | 70 | ||
NM_001259755.1 | NP_001246684.1 | 9 - 10 | 2710 | 70 | ||
NM_001274670.1 | NP_001261599.1 | 11 - 12 | 5649 | 70 | ||
NM_168293.3 | NP_729410.1 | 9 - 10 | 1708 | 70 | ||
NM_168294.3 | NP_729411.1 | 9 - 10 | 1708 | 70 | ||
GenBank mRNA | BT050456.1 | - | 9 - 10 | 1674 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |