Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169923.3 | NP_001163394.1 | 9 - 10 | 1412 | 103 | |
NM_168319.2 | NP_729457.1 | 8 - 9 | 1190 | 103 | ||
NM_168320.2 | NP_729458.1 | 8 - 9 | 1683 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |