Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bru3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202161.2 | NP_001189090.1 | 4 - 5 | 910 | 80 | |
NM_001202162.2 | NP_001189091.1 | 3 - 4 | 868 | 80 | ||
NM_001202163.2 | NP_001189092.1 | 3 - 4 | 868 | 80 | ||
NM_001202165.2 | NP_001189094.1 | 3 - 4 | 868 | 80 | ||
NM_001202166.4 | NP_001189095.1 | 3 - 4 | 868 | 80 | ||
NM_001274878.1 | NP_001261807.1 | 5 - 6 | 1252 | 80 | ||
NM_001274879.1 | NP_001261808.1 | 5 - 6 | 988 | 80 | ||
NM_001382115.1 | NP_001369049.1 | 3 - 4 | 868 | 80 | ||
NM_001382116.1 | NP_001369050.1 | 3 - 4 | 868 | 80 | ||
NM_001382117.1 | NP_001369051.1 | 4 - 5 | 853 | 80 | ||
NM_168557.3 | NP_729923.1 | 4 - 5 | 910 | 80 | ||
GenBank mRNA | AY060398.1 | - | 4 - 5 | 910 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |