Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104144.3 | NP_001097614.1 | 5 - 6 | 1504 | 69 | |
NM_001104145.3 | NP_001097615.1 | 5 - 6 | 1500 | 69 | ||
NM_001259854.2 | NP_001246783.1 | 5 - 6 | 1754 | 69 | ||
NM_001259855.2 | NP_001246784.1 | 5 - 6 | 1528 | 69 | ||
NM_001259856.2 | NP_001246785.1 | 5 - 6 | 1781 | 69 | ||
NM_001259857.2 | NP_001246786.1 | 5 - 6 | 1528 | 69 | ||
NM_001274990.1 | NP_001261919.1 | 5 - 6 | 1504 | 69 | ||
NM_001382059.1 | NP_001369057.1 | 5 - 6 | 1504 | 69 | ||
NM_168646.3 | NP_730099.2 | 5 - 6 | 1497 | 69 | ||
NM_168648.3 | NP_730101.2 | 5 - 6 | 1479 | 69 | ||
GenBank mRNA | BT133053.1 | - | 5 - 6 | 1485 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |