Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272502.1 | NP_001259431.1 | 3 - 4 | 1803 | 66 | |
NM_132421.3 | NP_572649.1 | 3 - 4 | 1838 | 66 | ||
GenBank mRNA | AY069308.1 | - | 3 - 4 | 1827 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |