Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169244.1 | NP_001162715.1 | 5 - 6 | 2109 | 145 | |
NM_001272500.1 | NP_001259429.1 | 4 - 5 | 1941 | 145 | ||
NM_132423.2 | NP_572651.1 | 3 - 4 | 1594 | 145 | ||
GenBank mRNA | BT015226.1 | - | 5 - 6 | 2109 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |