Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FucT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132512.4 | NP_572740.1 | 2 - 3 | 1295 | 89 | |
GenBank mRNA | AF441264.1 | - | 2 - 3 | 791 | 89 | |
AY051451.1 | - | 2 - 3 | 1246 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |