Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298397.1 | NP_001285326.1 | 1 - 2 | 145 | 56 | |
NM_132895.3 | NP_573123.1 | 1 - 2 | 145 | 56 | ||
GenBank mRNA | AJ238377.1 | - | 1 - 2 | 114 | 56 | |
AJ238378.1 | - | 1 - 2 | 111 | 56 | ||
AY051957.1 | - | 1 - 2 | 117 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |