Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273142.1 | NP_001260071.1 | 5 - 6 | 3099 | 61 | |
NM_001273143.1 | NP_001260072.1 | 5 - 6 | 3174 | 61 | ||
NM_057842.3 | NP_477190.1 | 5 - 6 | 3120 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |