Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cato | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057996.5 | NP_477344.1 | 1 | 552 | 92 | |
GenBank mRNA | BT028836.2 | - | 1 | 514 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |