Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080523.4 | NP_536784.1 | 2 - 3 | 287 | 118 | |
GenBank mRNA | AF233526.1 | - | 2 - 3 | 239 | 118 | |
AY051959.1 | - | 2 - 3 | 265 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |