Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170158.3 | NP_001163629.2 | 5 - 6 | 1156 | 76 | |
NM_001260211.2 | NP_001247140.1 | 5 - 6 | 1178 | 76 | ||
NM_057400.4 | NP_476748.1 | 5 - 6 | 1193 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |