Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276214.1 | NP_001263143.1 | 8 - 9 | 2511 | 84 | |
NM_057519.4 | NP_476867.1 | 9 - 10 | 2559 | 84 | ||
GenBank mRNA | BT031258.1 | - | 9 - 10 | 2023 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |