Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260338.2 | NP_001247267.1 | 3 - 4 | 899 | 144 | |
NM_057796.4 | NP_477144.1 | 3 - 4 | 899 | 144 | ||
GenBank mRNA | AF007805.1 | - | 3 - 4 | 884 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |