Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104217.2 | NP_001097687.1 | 2 - 3 | 843 | 80 | |
NM_079511.5 | NP_524235.2 | 3 - 4 | 1029 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |