Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-Man-IIa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275484.1 | NP_001262413.1 | 3 - 4 | 2264 | 72 | |
NM_079567.3 | NP_524291.2 | 3 - 4 | 2261 | 72 | ||
GenBank mRNA | AY119464.1 | - | 3 - 4 | 2194 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |