Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079649.4 | NP_524373.1 | 6 - 7 | 2588 | 93 | |
GenBank mRNA | AF071502.1 | - | 6 - 7 | 2487 | 93 | |
BT031130.1 | - | 6 - 7 | 2533 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |