Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079669.3 | NP_524393.2 | 11 - 12 | 1668 | 65 | |
NM_169777.2 | NP_732265.1 | 11 - 12 | 1738 | 65 | ||
GenBank mRNA | BT128700.1 | - | 11 - 12 | 1723 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |