Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141212.4 | NP_649469.2 | 4 - 5 | 662 | 73 | |
NM_168996.5 | NP_730835.2 | 3 - 4 | 588 | 73 | ||
GenBank mRNA | BT133109.1 | - | 3 - 4 | 585 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |