Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300242.1 | NP_001287171.1 | 2 - 3 | 1364 | 87 | |
NM_141268.3 | NP_649525.1 | 2 - 3 | 978 | 87 | ||
NM_169036.2 | NP_730906.1 | 2 - 3 | 1364 | 87 | ||
NM_169037.3 | NP_730908.1 | 2 - 3 | 543 | 87 | ||
GenBank mRNA | AY051594.1 | - | 2 - 3 | 609 | 87 | |
AY094896.1 | - | 2 - 3 | 1360 | 87 | ||
BT001659.1 | - | 2 - 3 | 538 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |