Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104224.2 | NP_001097694.1 | 3 - 4 | 578 | 56 | |
NM_141354.3 | NP_649611.2 | 3 - 4 | 578 | 56 | ||
GenBank mRNA | AY069336.1 | - | 3 - 4 | 546 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |