Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmdyn-D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141669.3 | NP_649926.2 | 2 - 3 | 431 | 73 | |
GenBank mRNA | BT150178.1 | - | 2 - 3 | 429 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |