Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Desat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141944.3 | NP_650201.1 | 1 - 2 | 423 | 73 | |
GenBank mRNA | BT024427.1 | - | 1 - 2 | 423 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |